5-Chloro-2-(4-methyl-1-piperidinyl)aniline

Supplier: Matrix Scientific

051127-500MG 051127-2.5G
101894-400EA 166.15 USD
101894-400 101894-402
5-Chloro-2-(4-methyl-1-piperidinyl)aniline
5-Chloro-2-(4-methyl-1-piperidinyl)aniline

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂
MDL Number: MFCD07365172
CAS Number: 893751-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR