5-Chloro-2-(2-methyl-1-piperidinyl)aniline

Supplier: Matrix Scientific

051125-500MG 051125-2.5G
101894-392EA 364.62 USD
101894-392 101894-394
5-Chloro-2-(2-methyl-1-piperidinyl)aniline
5-Chloro-2-(2-methyl-1-piperidinyl)aniline

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂
MDL Number: MFCD07365170
CAS Number: 893751-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR