2-(2-Amino-4-chloroethylanilino)-1-ethanol

Supplier: Matrix Scientific

051123-500MG 051123-2.5G
101894-384EA 364.62 USD
101894-384 101894-386
2-(2-Amino-4-chloroethylanilino)-1-ethanol
2-(2-Amino-4-chloroethylanilino)-1-ethanol

Hazard: Irritant

Formula: C₁₀H₁₅ClN₂O
MDL Number: MFCD12772395
CAS Number: 1179262-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR