2-(1-Azepanyl)-3-chloroaniline
Supplier: Matrix Scientific
051103-2.5G
051103-500MG
101894-304EA
364.62
USD
101894-304
101894-306
2-(1-Azepanyl)-3-chloroaniline
2-(1-Azepanyl)-3-chloroaniline
Hazard: Irritant
Formula:
C₁₂H₁₇ClN₂ |
MDL Number:
MFCD08700273 CAS Number: 926215-35-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...