2-(2-Amino-6-chloroethylanilino)-1-ethanol

Supplier: Matrix Scientific

051098-500MG 051098-2.5G
101894-284EA 364.62 USD
101894-284 101894-286
2-(2-Amino-6-chloroethylanilino)-1-ethanol
2-(2-Amino-6-chloroethylanilino)-1-ethanol

Hazard: Irritant

Formula: C₁₀H₁₅ClN₂O
MDL Number: MFCD12813170
CAS Number: 1178691-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR