2-[3,4-Dihydro-1(2H)-quinolinyl]aniline
Supplier: Matrix Scientific
051090-2.5G
051090-500MG
101894-252EA
364.62
USD
101894-252
101894-254
2-[3,4-Dihydro-1(2H)-quinolinyl]aniline
2-[3,4-Dihydro-1(2H)-quinolinyl]aniline
Hazard: Irritant
Formula:
C₁₅H₁₆N₂ |
MDL Number:
MFCD08699290 CAS Number: 112631-31-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...