2-(2,3-Dihydro-1H-indol-1-yl)phenylamine

Supplier: Matrix Scientific

051089-2.5G 051089-500MG
101894-250EA 838.46 USD
101894-250 101894-248
2-(2,3-Dihydro-1H-indol-1-yl)phenylamine
2-(2,3-Dihydro-1H-indol-1-yl)phenylamine

Hazard: Irritant

Formula: C₁₄H₁₄N₂
MDL Number: MFCD08699289
CAS Number: 180629-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR