1-(2-Aminophenyl)-4-piperidinol

Supplier: Matrix Scientific

051082-500MG 051082-2.5G
101894-220EA 272.55 USD
101894-220 101894-222
1-(2-Aminophenyl)-4-piperidinol
1-(2-Aminophenyl)-4-piperidinol

Hazard: Irritant

Formula: C₁₁H₁₆N₂O
MDL Number: MFCD09734091
CAS Number: 252758-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR