Methyl (2S,4S)-4-(2-bromo-4-fluorophenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051060-500MG 051060-2.5G
101894-132EA 364.62 USD
101894-132 101894-134
Methyl (2S,4S)-4-(2-bromo-4-fluorophenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(2-bromo-4-fluorophenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₄BrClFNO₃
MDL Number: MFCD13561429
CAS Number: 1354488-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR