Methyl (2S,4S)-4-[(4-chloro-1-naphthyl)oxy]-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051055-500MG 051055-2.5G
101894-112EA 364.62 USD
101894-112 101894-114
Methyl (2S,4S)-4-[(4-chloro-1-naphthyl)oxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[(4-chloro-1-naphthyl)oxy]-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₆H₁₇Cl₂NO₃
MDL Number: MFCD13561424
CAS Number: 1354488-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR