Methyl (2S,4S)-4-(2-bromo-4-chlorophenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051041-500MG 051041-2.5G
101894-056EA 364.62 USD
101894-056 101894-058
Methyl (2S,4S)-4-(2-bromo-4-chlorophenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(2-bromo-4-chlorophenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₄BrCl₂NO₃
MDL Number: MFCD13561411
CAS Number: 1354487-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR