Methyl (2S,4S)-4-(4-bromo-2-chlorophenoxy)-2-pyrrolidinecarboxylate hydrochloride
Supplier: Matrix Scientific
051022-500MG
051022-2.5G
101893-982EA
987.79
USD
101893-982
101893-980
Methyl (2S,4S)-4-(4-bromo-2-chlorophenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(4-bromo-2-chlorophenoxy)-2-pyrrolidinecarboxylate hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₄BrCl₂NO₃ |
MDL Number:
MFCD13561392 CAS Number: 1354487-92-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...