(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2,4-dibromophenoxy)-2-pyrrolidinecarboxylic acid
Supplier: Matrix Scientific
051020-2.5G
051020-500MG
101893-972EA
364.62
USD
101893-972
101893-974
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2,4-dibromophenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2,4-dibromophenoxy)-2-pyrrolidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₆H₁₉Br₂NO₅ |
MDL Number:
MFCD13561390 CAS Number: 1354487-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...