Methyl (2S,4S)-4-(2-bromophenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051019-500MG 051019-2.5G
101893-970EA 838.46 USD
101893-970 101893-968
Methyl (2S,4S)-4-(2-bromophenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(2-bromophenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₅BrClNO₃
MDL Number: MFCD13561389
CAS Number: 1354487-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR