(2S,4S)-4-(2-Acetylphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
Supplier: Matrix Scientific
051013-500MG
051013-2.5G
101893-946EA
838.46
USD
101893-946
101893-944
(2S,4S)-4-(2-Acetylphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
(2S,4S)-4-(2-Acetylphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₈H₂₃NO₆ |
MDL Number:
MFCD13561383 CAS Number: 1266111-72-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...