Methyl (2S,4S)-4-[4-(ethoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Supplier: Matrix Scientific
051006-500MG
051006-2.5G
101893-918EA
1896.13
USD
101893-918
101893-916
Methyl (2S,4S)-4-[4-(ethoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[4-(ethoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₀ClNO₅ |
MDL Number:
MFCD13561376 CAS Number: 1354487-46-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...