Methyl (2S,4S)-4-[2-(methoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051004-500MG 051004-2.5G
101893-908EA 364.62 USD
101893-908 101893-910
Methyl (2S,4S)-4-[2-(methoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[2-(methoxycarbonyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₈ClNO₅
MDL Number: MFCD13561374
CAS Number: 1354486-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR