Methyl (2S,4S)-4-[3-(diethylamino)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Supplier: Matrix Scientific
051002-2.5G
051002-500MG
101893-902EA
838.46
USD
101893-902
101893-900
Methyl (2S,4S)-4-[3-(diethylamino)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[3-(diethylamino)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Hazard: Irritant
Formula:
C₁₆H₂₅ClN₂O₃ |
MDL Number:
MFCD13561372 CAS Number: 1354484-76-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...