Methyl (2S,4S)-4-[2-chloro-4-(tert-pentyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Supplier: Matrix Scientific
050984-2.5G
050984-500MG
101893-828EA
364.62
USD
101893-828
101893-830
Methyl (2S,4S)-4-[2-chloro-4-(tert-pentyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[2-chloro-4-(tert-pentyl)phenoxy]-2-pyrrolidinecarboxylate hydrochloride
Hazard: Irritant
Formula:
C₁₇H₂₅Cl₂NO₃ |
MDL Number:
MFCD13561354 CAS Number: 1354485-03-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...