(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-chloro-2-ethylphenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

050979-2.5G 050979-500MG
101893-808EA 272.55 USD
101893-808 101893-810
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-chloro-2-ethylphenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-chloro-2-ethylphenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₈H₂₄ClNO₅
MDL Number: MFCD13561349
CAS Number: 1354486-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR