(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-chloro-2-isopropylphenoxy)-2-pyrrolidinecarboxylic acid
Supplier: Matrix Scientific
050975-2.5G
050975-500MG
101893-792EA
364.62
USD
101893-792
101893-794
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-chloro-2-isopropylphenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(4-chloro-2-isopropylphenoxy)-2-pyrrolidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₉H₂₆ClNO₅ |
MDL Number:
MFCD13561345 CAS Number: 1354486-29-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...