(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-chloro-4-isopropylphenoxy)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

050967-500MG 050967-2.5G
101893-762EA 838.46 USD
101893-762 101893-760
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-chloro-4-isopropylphenoxy)-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-(2-chloro-4-isopropylphenoxy)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₉H₂₆ClNO₅
MDL Number: MFCD13561337
CAS Number: 1354486-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR