(2S,4S)-4-[(5-Bromo[1,1'-biphenyl]-2-yl)oxy]-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

050959-500MG 050959-2.5G
101893-728EA 364.62 USD
101893-728 101893-730
(2S,4S)-4-[(5-Bromo[1,1'-biphenyl]-2-yl)oxy]-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
(2S,4S)-4-[(5-Bromo[1,1'-biphenyl]-2-yl)oxy]-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₂₂H₂₄BrNO₅
MDL Number: MFCD13561329
CAS Number: 1354487-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR