(2S,4S)-4-(2-Bromo-4-ethylphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
Supplier: Matrix Scientific
050951-500MG
050951-2.5G
101893-696EA
364.62
USD
101893-696
101893-698
(2S,4S)-4-(2-Bromo-4-ethylphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
(2S,4S)-4-(2-Bromo-4-ethylphenoxy)-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₈H₂₄BrNO₅ |
MDL Number:
MFCD13561321 CAS Number: 1354486-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...