(2S,4S)-4-[2-Bromo-4-(tert-butyl)phenoxy]-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
Supplier: Matrix Scientific
050949-2.5G
050949-500MG
101893-690EA
838.46
USD
101893-690
101893-688
(2S,4S)-4-[2-Bromo-4-(tert-butyl)phenoxy]-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
(2S,4S)-4-[2-Bromo-4-(tert-butyl)phenoxy]-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
Hazard: Irritant
Formula:
C₂₀H₂₈BrNO₅ |
MDL Number:
MFCD13561319 CAS Number: 1354484-68-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...