Methyl (2S,4S)-4-(2-bromo-4-isopropylphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Supplier: Matrix Scientific
050946-2.5G
050946-500MG
101893-678EA
838.46
USD
101893-678
101893-676
Methyl (2S,4S)-4-(2-bromo-4-isopropylphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(2-bromo-4-isopropylphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₁BrClNO₃ |
MDL Number:
MFCD13561316 CAS Number: 1354486-46-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...