(2S,4S)-4-[4-Bromo-2-(tert-butyl)phenoxy]-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Supplier: Matrix Scientific

050941-500MG 050941-2.5G
101893-656EA 364.62 USD
101893-656 101893-658
(2S,4S)-4-[4-Bromo-2-(tert-butyl)phenoxy]-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid
(2S,4S)-4-[4-Bromo-2-(tert-butyl)phenoxy]-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₂₀H₂₈BrNO₅
MDL Number: MFCD13561311
CAS Number: 1354485-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR