Methyl (2S,4S)-4-(4-bromo-2-isopropylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

050940-500MG 050940-2.5G
101893-652EA 364.62 USD
101893-652 101893-654
Methyl (2S,4S)-4-(4-bromo-2-isopropylphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(4-bromo-2-isopropylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₁BrClNO₃
MDL Number: MFCD13561310
CAS Number: 1354486-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR