(2S,4S)-1-(tert-Butoxycarbonyl)-4-[(2,4-dichloro-1-naphthyl)oxy]-2-pyrrolidine carboxylic acid

Supplier: Matrix Scientific

050933-500MG 050933-2.5G
101893-624EA 364.62 USD
101893-624 101893-626
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[(2,4-dichloro-1-naphthyl)oxy]-2-pyrrolidine carboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-[(2,4-dichloro-1-naphthyl)oxy]-2-pyrrolidine carboxylic acid

Hazard: Irritant

Formula: C₂₀H₂₁Cl₂NO₅
MDL Number: MFCD13561303
CAS Number: 1354485-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR