4-(Isopropoxymethyl)piperidine hydrochloride

Supplier: Matrix Scientific

050911-2.5G 050911-500MG
101893-536EA 364.62 USD
101893-536 101893-538
4-(Isopropoxymethyl)piperidine hydrochloride
4-(Isopropoxymethyl)piperidine hydrochloride

Hazard: Irritant

Formula: C₉H₂₀ClNO
MDL Number: MFCD13561281
CAS Number: 1211439-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR