2-(3-Pyrrolidinyloxy)-5-(trifluoromethyl)pyridine hydrochloride

Supplier: Matrix Scientific

050897-500MG 050897-2.5G
101893-484EA 124.2 USD
101893-484 101893-486
2-(3-Pyrrolidinyloxy)-5-(trifluoromethyl)pyridine hydrochloride
2-(3-Pyrrolidinyloxy)-5-(trifluoromethyl)pyridine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₂ClF₃N₂O
MDL Number: MFCD13561268
CAS Number: 1220016-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR