2-(4-Piperidinylmethoxy)-5-(trifluoromethyl)pyridine hydrochloride

Supplier: Matrix Scientific

050896-2.5G 050896-500MG
101893-482EA 838.46 USD
101893-482 101893-480
2-(4-Piperidinylmethoxy)-5-(trifluoromethyl)pyridine hydrochloride
2-(4-Piperidinylmethoxy)-5-(trifluoromethyl)pyridine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆ClF₃N₂O
MDL Number: MFCD13561267
CAS Number: 1220036-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR