4-Chloro-6-(4-piperidinylmethoxy)pyrimidine hydrochloride

Supplier: Matrix Scientific

050876-2.5G 050876-500MG
101893-400EA 364.62 USD
101893-400 101893-402
4-Chloro-6-(4-piperidinylmethoxy)pyrimidine hydrochloride
4-Chloro-6-(4-piperidinylmethoxy)pyrimidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₅Cl₂N₃O
MDL Number: MFCD13561253
CAS Number: 1220036-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR