6-Chloro-4-pyrimidinyl-3-piperidinyl ether hydrochloride
Supplier: Matrix Scientific
050873-2.5G
050873-500MG
101893-388EA
364.62
USD
101893-388
101893-390
6-Chloro-4-pyrimidinyl-3-piperidinyl ether hydrochloride
6-Chloro-4-pyrimidinyl-3-piperidinyl ether hydrochloride
Hazard: Irritant
Formula:
C₉H₁₃Cl₂N₃O MW: 250.13 g/mol |
MDL Number:
MFCD09607627 CAS Number: 1185311-81-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...