4-[(3,5-Dimethoxybenzyl)oxy]piperidine hydrochloride

Supplier: Matrix Scientific

050864-500MG 050864-2.5G
101893-352EA 364.62 USD
101893-352 101893-354
4-[(3,5-Dimethoxybenzyl)oxy]piperidine hydrochloride
4-[(3,5-Dimethoxybenzyl)oxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClNO₃
MDL Number: MFCD13561244
CAS Number: 1220036-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR