4-[(2,4-Difluorobenzyl)oxy]piperidine hydrochloride
Supplier: Matrix Scientific
050844-2.5G
050844-500MG
101893-274EA
568.57
USD
101893-274
101893-272
4-[(2,4-Difluorobenzyl)oxy]piperidine hydrochloride
4-[(2,4-Difluorobenzyl)oxy]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₆ClF₂NO |
MDL Number:
MFCD13561224 CAS Number: 1220034-23-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...