Methyl-4-[(3-pyrrolidinyloxy)methyl]benzoate hydrochloride
Supplier: Matrix Scientific
050827-500MG
050827-2.5G
101893-206EA
838.46
USD
101893-206
101893-204
Methyl-4-[(3-pyrrolidinyloxy)methyl]benzoate hydrochloride
Methyl-4-[(3-pyrrolidinyloxy)methyl]benzoate hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₈ClNO₃ |
MDL Number:
MFCD13561207 CAS Number: 1220034-80-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...