4-[(2-Methoxy-4-propylphenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050816-500MG 050816-2.5G
101893-162EA 838.46 USD
101893-162 101893-160
4-[(2-Methoxy-4-propylphenoxy)methyl]piperidine hydrochloride
4-[(2-Methoxy-4-propylphenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆ClNO₂
MDL Number: MFCD13561196
CAS Number: 1219977-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR