4-[(2-Iodophenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050806-500MG 050806-2.5G
101893-120EA 272.55 USD
101893-120 101893-122
4-[(2-Iodophenoxy)methyl]piperidine hydrochloride
4-[(2-Iodophenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClINO
MDL Number: MFCD13561186
CAS Number: 1220034-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR