4-[(2-Nitrophenoxy)methyl]piperidine hydrochloride
Supplier: Matrix Scientific
050801-2.5G
050801-500MG
101893-102EA
838.46
USD
101893-102
101893-100
4-[(2-Nitrophenoxy)methyl]piperidine hydrochloride
4-[(2-Nitrophenoxy)methyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₇ClN₂O₃ |
MDL Number:
MFCD12407034 CAS Number: 614730-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...