2-[(4-Aminophenyl)sulfanyl]-N-(2,5-difluorophenyl)acetamide ≥95%

Supplier: Matrix Scientific

047686-500MG 047686-1G
101880-984EA 181.54 USD
101880-984 101880-986
2-[(4-Aminophenyl)sulfanyl]-N-(2,5-difluorophenyl)acetamide ≥95%
2-[(4-Aminophenyl)sulfanyl]-N-(2,5-difluorophenyl)acetamide

Hazard: Irritant

Formula: C₁₄H₁₂F₂N₂OS
MW: 294.33 g/mol
MDL Number: MFCD07021305
CAS Number: 885267-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR