2-[(4-Aminophenyl)sulfanyl]-N-[4-(trifluoromethoxy)phenyl]acetamide ≥95%
Supplier: Matrix Scientific
047685-1G
047685-500MG
101880-980EA
181.54
USD
101880-980
101880-982
2-[(4-Aminophenyl)sulfanyl]-N-[4-(trifluoromethoxy)phenyl]acetamide ≥95%
2-[(4-Aminophenyl)sulfanyl]-N-[4-(trifluoromethoxy)phenyl]acetamide
Hazard: Irritant
Formula:
C₁₅H₁₃F₃N₂O₂S |
MDL Number:
MFCD05155029 CAS Number: 763126-91-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...