2-[(4-Aminophenyl)sulfanyl]-N-(4-butylphenyl)acetamide ≥95%

Supplier: Matrix Scientific

047684-500MG
101880-978EA 181.54 USD
101880-978
2-[(4-Aminophenyl)sulfanyl]-N-(4-butylphenyl)acetamide ≥95%
2-[(4-Aminophenyl)sulfanyl]-N-(4-butylphenyl)acetamide

Hazard: Irritant

Formula: C₁₈H₂₂N₂OS
MW: 314.45 g/mol
MDL Number: MFCD05155026
CAS Number: 763128-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR