4-Butyl-5-phenyl-2,4-dihydro-3H-1,2,4-triazol-3-one ≥95%

Supplier: Matrix Scientific

047683-1G 047683-500MG
101880-974EA 135.7 USD
101880-974 101880-976
4-Butyl-5-phenyl-2,4-dihydro-3H-1,2,4-triazol-3-one ≥95%
4-Butyl-5-phenyl-2,4-dihydro-3H-1,2,4-triazol-3-one

Hazard: Irritant

Formula: C₁₂H₁₅N₃O
MW: 217.27 g/mol
MDL Number: MFCD00098834
CAS Number: 271798-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR