5-[3-(2,4-Dichlorophenoxy)propyl]-4-phenyl-4H-1,2,4-triazole-3-thiol ≥95%

Supplier: Matrix Scientific

047670-500MG
101880-926EA 181.54 USD
101880-926
5-[3-(2,4-Dichlorophenoxy)propyl]-4-phenyl-4H-1,2,4-triazole-3-thiol ≥95%
5-[3-(2,4-Dichlorophenoxy)propyl]-4-phenyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₇H₁₅Cl₂N₃OS
MW: 380.3 g/mol
MDL Number: MFCD05150562
CAS Number: 379704-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR