DL-6-Hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Hydroxy-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid

047459-1G
101880-100EA 310.5 USD
101880-100
DL-6-Hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate ≥95%
DL-6-Hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrate

Hazard: Irritant

Formula: C₁₀H₁₁NO₃.xH₂O
MW: 193.2 g/mol (anhy)
MDL Number: MFCD00800582
CAS Number: 76824-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR