Methyl-2-chloro-4-(trifluoromethyl)nicotinate ≥95%

Supplier: Matrix Scientific

047431-500MG
101879-944EA 410.55 USD
101879-944
Methyl-2-chloro-4-(trifluoromethyl)nicotinate ≥95%
Methyl-2-chloro-4-(trifluoromethyl)nicotinate

Hazard: Irritant

Formula: C₈H₅ClF₃NO₂
MW: 239.58 g/mol
MDL Number: MFCD12827815
CAS Number: 196708-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR