(2-Chloro-3-pyridinyl)-N-methylmethanamine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-3-[(methylamino)methyl]pyridine

047423-500MG
101879-916EA 201.25 USD
101879-916
(2-Chloro-3-pyridinyl)-N-methylmethanamine ≥95%
(2-Chloro-3-pyridinyl)-N-methylmethanamine

Hazard: Irritant

Formula: C₇H₉ClN₂
MW: 156.62 g/mol
MDL Number: MFCD09864374
CAS Number: 120739-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR