Ethyl imidazo[1,2-a]pyridine-8-carboxylate ≥95%

Supplier: Matrix Scientific

047422-500MG
101879-912EA 356.92 USD
101879-912
Ethyl imidazo[1,2-a]pyridine-8-carboxylate ≥95%
Ethyl imidazo[1,2-a]pyridine-8-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₂
MDL Number: MFCD10698079
CAS Number: 158020-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR