Ethyl-3-(chlorosulfonyl)propanoate ≥90%

Supplier: Matrix Scientific

047391-1G
101879-786EA 623.08 USD
101879-786
Ethyl-3-(chlorosulfonyl)propanoate ≥90%
Ethyl-3-(chlorosulfonyl)propanoate

Hazard: Refrigerate/Stored under Argon

Formula: C₅H₉ClO₄S
MW: 200.64 g/mol
MDL Number: MFCD07186267
CAS Number: 103472-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR